accessibility menu, dialog, popup

UserName

HOBrHO

CAS RN 2316-64-5

IUPAC Name: 4-bromo-2-(hydroxymethyl)phenol
Synonyms: 4-bromo-2-(hydroxymethyl)phenol
Molecular Weight (g/mol): 203.04
Molecular Formula: C7H7BrO2
InChi Key: KNKRHSVKIORZQB-UHFFFAOYSA-N
SMILES: OCC1=CC(Br)=CC=C1O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results