accessibility menu, dialog, popup

UserName

CAS RN 2259-96-3

IUPAC Name: 3-{bicyclo[2.2.1]hept-5-en-2-yl}-6-chloro-1,1-dioxo-3,4-dihydro-2H-1λ⁶,2,4-benzothiadiazine-7-sulfonamide
Synonyms: cyclothiazide fluidil renazide doburil cyclothiazide preparation cyclothiazide (substance) 环噻嗪 cyclothiazidum aquirel anhydron
Molecular Weight (g/mol): 389.87
Molecular Formula: C14H16ClN3O4S2
InChi Key: BOCUKUHCLICSIY-UHFFFAOYNA-N
SMILES: NS(=O)(=O)C1=CC2=C(NC(NS2(=O)=O)C2CC3CC2C=C3)C=C1Cl
Certifications/Compliance 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)