CAS RN 22224-41-5
IUPAC Name:
[3,5-bis(benzoyloxy)-4-hydroxyoxolan-2-yl]methyl benzoate
Synonyms:
[3,5-bis(benzoyloxy)-4-hydroxyoxolan-2-yl]methyl benzoate
Molecular Weight (g/mol):
462.45
Molecular Formula:
C26H22O8
InChi Key:
HUHVPBKTTFVAQF-UHFFFAOYNA-N
SMILES:
OC1C(OC(=O)C2=CC=CC=C2)OC(COC(=O)C2=CC=CC=C2)C1OC(=O)C1=CC=CC=C1
1
–
3
of
3
results
1
1,3,5-Tri-O-benzoyl-alpha-D-ribofuranose, 97%, Thermo Scientific Chemicals
CAS: 22224-41-5 Molecular Formula: C26H22O8 Molecular Weight (g/mol): 462.454 MDL Number: MFCD00080818 InChI Key: HUHVPBKTTFVAQF-GCUWJAMBSA-N Synonym: α-D-Ribofuranose 1,3,5-tribenzoate PubChem CID: 68022796 IUPAC Name: [(2R,5R)-3,5-dibenzoyloxy-4-hydroxyoxolan-2-yl]methyl benzoate SMILES: C1=CC=C(C=C1)C(=O)OCC2C(C(C(O2)OC(=O)C3=CC=CC=C3)O)OC(=O)C4=CC=CC=C4
3
1,3,5-Tri-O-benzoyl-a-D-ribofuranose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.