missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tri-O-Benzoyl-α-D-Ribofuranose, MP Biomedicals™

Product Code. 11437422 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11437422 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11437422 Supplier MP Biomedicals Supplier No. 0215396801
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 22224-41-5
Color White
Molecular Formula C26H22O8
Quantity 1 g
Synonym α-D-Ribofuranose 1, 3, 5-tribenzoate
InChI Key HUHVPBKTTFVAQF-GCUWJAMBSA-N
SMILES C1=CC=C(C=C1)C(=O)OCC2C(C(C(O2)OC(=O)C3=CC=CC=C3)O)OC(=O)C4=CC=CC=C4
IUPAC Name [(2R,5R)-3,5-dibenzoyloxy-4-hydroxyoxolan-2-yl]methyl benzoate
Molecular Weight (g/mol) 462.454
PubChem CID 68022796
Formula Weight 462.4
Physical Form Powder
Chemical Name or Material 1, 3, 5-Tri-O-Benzoyl-α-D-Ribofuranose
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.