accessibility menu, dialog, popup

UserName

ON

CAS RN 22065-85-6

IUPAC Name: 1-benzylpiperidine-4-carbaldehyde
Synonyms: 1-benzylpiperidine-4-carbaldehyde
Molecular Weight (g/mol): 203.29
Molecular Formula: C13H17NO
InChi Key: SGIBOXBBPQRZDM-UHFFFAOYSA-N
SMILES: O=CC1CCN(CC2=CC=CC=C2)CC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results