accessibility menu, dialog, popup

UserName

CAS RN 219539-29-4

H3CONSCl

CAS RN 219539-29-4

IUPAC Name: 1-[2-(4-chlorophenyl)-6H-1,3-thiazin-5-yl]ethan-1-one
Synonyms: 1-[2-(4-chlorophenyl)-6H-1,3-thiazin-5-yl]ethanone
Molecular Weight (g/mol): 251.73
Molecular Formula: C12H10ClNOS
InChi Key: NEKOTKLWPGNIOI-UHFFFAOYSA-N
SMILES: CC(=O)C1=CN=C(SC1)C1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results