accessibility menu, dialog, popup

UserName

CH3CH3CH3OOHNOOOOH

CAS RN 218943-30-7

IUPAC Name: 6-(benzyloxy)-3-{[(tert-butoxy)carbonyl]amino}-6-oxohexanoic acid
Synonyms: 6-(benzyloxy)-3-[(tert-butoxycarbonyl)amino]-6-oxohexanoic acid
Molecular Weight (g/mol): 351.4
Molecular Formula: C18H25NO6
InChi Key: JABNOCWCLLYHKE-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CCC(=O)OCC1=CC=CC=C1)CC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results