CAS RN 21581-49-7
CAS RN 21581-49-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6-Nitrodopamine, TRC
CAS: 21581-49-7 Molecular Formula: C8H10N2O4 Molecular Weight (g/mol): 198.18 Synonym: 4-(2-Aminoethyl)-5-nitropyrocatechol,4-(2-Aminoethyl)-5-nitro-1,2-benzenediol,4-(2-Aminoethyl)-5-nitrobenzene-1,2-diol SMILES: CC1c2cc3C(C)(C)C(C)C(C)(C)c3cc2COC1