CAS RN 2113-05-5
CAS RN 2113-05-5
IUPAC Name:
N'-(azaniumylmethanimidoyl)-N-(3-chlorophenyl)guanidine chloride
Synonyms:
N'-(ammoniomethanimidoyl)-N-(3-chlorophenyl)guanidine chloride
Molecular Weight (g/mol):
248.11
Molecular Formula:
C8H11Cl2N5
InChi Key:
FOWAIJYHRWFTHR-UHFFFAOYSA-N
SMILES:
[Cl-].[NH3+]C(=N)NC(=N)NC1=CC(Cl)=CC=C1
1
–
2
of
2
results
1
1-(3-Chlorophenyl)biguanide hydrochloride, 97%
CAS: 2113-05-5 MDL Number: MFCD00053019