CAS RN 21036-68-0
IUPAC Name:
2-hydrazinyl-4-(thiophen-2-yl)-1,3-thiazole
Synonyms:
2-hydrazinyl-4-(thiophen-2-yl)-1,3-thiazole
Molecular Weight (g/mol):
197.27
Molecular Formula:
C7H7N3S2
InChi Key:
QYFFQQHJTLJVGU-UHFFFAOYSA-N
SMILES:
NNC1=NC(=CS1)C1=CC=CS1
1
–
1
of
1
results
1
2-Hydrazino-4-(2-thienyl)thiazole, 97%, Thermo Scientific™
CAS: 21036-68-0 Molecular Formula: C7H7N3S2 Molecular Weight (g/mol): 197.27 MDL Number: MFCD06739917 InChI Key: QYFFQQHJTLJVGU-UHFFFAOYSA-N Synonym: 4-thiophen-2-yl-thiazol-2-yl-hydrazine,2-hydrazinyl-4-thiophen-2-yl-1,3-thiazole,2-hydrazino-4-2-thienyl thiazole,4-thiophen-2-ylthiazol-2-yl hydrazine,2-hydrazinyl-4-thiophen-2-yl thiazole,4-thiophen-2-yl-1,3-thiazol-2-yl hydrazine PubChem CID: 7174026 SMILES: NNC1=NC(=CS1)C1=CC=CS1