accessibility menu, dialog, popup

UserName

FFFOBrBr

CAS RN 207226-31-1

IUPAC Name: 1,3-dibromo-5-(trifluoromethoxy)benzene
Synonyms: 1,3-dibromo-5-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 319.9
Molecular Formula: C7H3Br2F3O
InChi Key: UKHOUWWEEAOCTI-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC(Br)=CC(Br)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results