accessibility menu, dialog, popup

UserName

CAS RN 206996-13-6

CH3NNClClHClHCl

CAS RN 206996-13-6

IUPAC Name: 1-[2-(3,4-dichlorophenyl)ethyl]-4-methylpiperazine dihydrochloride
Synonyms: 1-[2-(3,4-dichlorophenyl)ethyl]-4-methylpiperazine dihydrochloride
Molecular Weight (g/mol): 346.12
Molecular Formula: C13H20Cl4N2
InChi Key: NXFDBTLQOARIMH-UHFFFAOYSA-N
SMILES: Cl.Cl.CN1CCN(CCC2=CC(Cl)=C(Cl)C=C2)CC1
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
Color 
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results