missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD 1063 Dihydrochloride, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | 1-[2-(3,4-Dichlorophenyl)ethyl]-4-methylpiperazine Dihydrochloride |
| CAS | 206996-13-6 |
| Molecular Formula | C13H20Cl4N2 |
| Quantity | 5 mg |
| Synonym | BD 1063 Dihydrochloride |
| InChI Formula | InChI=1S/C13H18Cl2N2.2ClH/c1-16-6-8-17(9-7-16)5-4-11-2-3-12(14)13(15)10-11;;/h2-3,10H,4-9H2,1H3;2*1H |
| SMILES | Cl.Cl.CN1CCN(CCc2ccc(Cl)c(Cl)c2)CC1 |
| IUPAC Name | 1-[2-(3,4-dichlorophenyl)ethyl]-4-methylpiperazine;dihydrochloride |
| Molecular Weight (g/mol) | 346.12 |
| Formula Weight | 344.0381 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction