accessibility menu, dialog, popup

UserName

CH3H3CN(R)OH(S)NHOO(R)O(R)O(S)SOONH2

CAS RN 206361-99-1

IUPAC Name: (3R,3aS,6aR)-hexahydrofuro[2,3-b]furan-3-yl N-[(2S,3R)-3-hydroxy-4-[N-(2-methylpropyl)4-aminobenzenesulfonamido]-1-phenylbutan-2-yl]carbamate
Synonyms: darunavir prezista darunavirum
Molecular Weight (g/mol): 547.67
Molecular Formula: C27H37N3O7S
InChi Key: CJBJHOAVZSMMDJ-HEXNFIEUSA-N
SMILES: CC(C)CN(C[C@@H](O)[C@H](CC1=CC=CC=C1)NC(=O)O[C@H]1CO[C@H]2OCC[C@@H]12)S(=O)(=O)C1=CC=C(N)C=C1
Molecular Weight (g/mol) 
  • (6)
  • (1)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Color 
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results