accessibility menu, dialog, popup

UserName

H3CCH3H3COONOOCH3FF

CAS RN 203866-17-5

IUPAC Name: 1-tert-butyl 2-methyl 4,4-difluoropyrrolidine-1,2-dicarboxylate
Synonyms: 1-tert-butyl 2-methyl 4,4-difluoropyrrolidine-1,2-dicarboxylate
Molecular Weight (g/mol): 265.26
Molecular Formula: C11H17F2NO4
InChi Key: RQDZKOOUQIDZOG-UHFFFAOYNA-N
SMILES: COC(=O)C1CC(F)(F)CN1C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results