accessibility menu, dialog, popup

UserName

CAS RN 2016-88-8

HClH2NNH2NONClNH2NH2N

CAS RN 2016-88-8

IUPAC Name: hydrogen 3,5-diamino-6-chloro-N-(diaminomethylidene)pyrazine-2-carboxamide chloride
Synonyms: amiloride
Molecular Weight (g/mol): 266.09
Molecular Formula: C6H9Cl2N7O
InChi Key: ACHKKGDWZVCSNH-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC(N)=NC(=O)C1=NC(Cl)=C(N)N=C1N
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)