CAS RN 200807-45-0
IUPAC Name:
2-chloro-N-(2,6-dichlorophenyl)benzamide
Synonyms:
2-chloro-N-(2,6-dichlorophenyl)benzamide
Molecular Weight (g/mol):
300.56
Molecular Formula:
C13H8Cl3NO
InChi Key:
MLILWLKTWNAOFL-UHFFFAOYSA-N
SMILES:
ClC1=CC=CC(Cl)=C1NC(=O)C1=CC=CC=C1Cl
1
–
1
of
1
results
1
2-Chloro-N-(2,6-dichlorophenyl)benzamide, 97%, Thermo Scientific™
CAS: 200807-45-0 Molecular Formula: C13H8Cl3NO Molecular Weight (g/mol): 300.563 MDL Number: MFCD00028398 InChI Key: MLILWLKTWNAOFL-UHFFFAOYSA-N Synonym: 2-chloro-n-2,6-dichlorophenyl benzamide,2,2',6'-trichlorobenzanilide PubChem CID: 853837 IUPAC Name: 2-chloro-N-(2,6-dichlorophenyl)benzamide SMILES: C1=CC=C(C(=C1)C(=O)NC2=C(C=CC=C2Cl)Cl)Cl