accessibility menu, dialog, popup

UserName

CH3O

CAS RN 19832-98-5

IUPAC Name: 4-methyl-1,2,3,4-tetrahydronaphthalen-1-one
Synonyms: 4-methyl-3,4-dihydro-2H-naphthalen-1-one
Molecular Weight (g/mol): 160.22
Molecular Formula: C11H12O
InChi Key: SRLHDEROUKFEMJ-UHFFFAOYNA-N
SMILES: CC1CCC(=O)C2=CC=CC=C12
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results