CAS RN 19780-71-3
IUPAC Name:
5-ethylnonan-3-ol
Synonyms:
5-ethylnonan-3-ol
Molecular Weight (g/mol):
172.31
Molecular Formula:
C11H24O
InChi Key:
DWMKGYOHPSYFCS-UHFFFAOYNA-N
SMILES:
CCCCC(CC)CC(O)CC
1
–
1
of
1
results
1
5-Ethyl-3-nonanol, erythro + threo, 99%, Thermo Scientific™
CAS: 19780-71-3 Molecular Formula: C11H24O Molecular Weight (g/mol): 172.312 MDL Number: MFCD00046677 InChI Key: DWMKGYOHPSYFCS-UHFFFAOYSA-N Synonym: 5-ethyl-3-nonanol,3-nonanol, 5-ethyl,acmc-1br0g,5-ethyl-3-nonanol, erythro + threo,5-ethyl-3-nonanol, threo + erythro PubChem CID: 519707 IUPAC Name: 5-ethylnonan-3-ol SMILES: CCCCC(CC)CC(CC)O