CAS RN 196309-76-9
CAS RN 196309-76-9
IUPAC Name:
3-(4-tert-butylbenzenesulfonyl)prop-2-enenitrile
Synonyms:
3-(4-tert-butylbenzenesulfonyl)prop-2-enenitrile
Molecular Weight (g/mol):
249.33
Molecular Formula:
C13H15NO2S
InChi Key:
VHKZGNPOHPFPER-UHFFFAOYSA-N
SMILES:
CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)C=CC#N
1
–
3
of
3
results
1
BAY 11-7085, TRC
CAS: 196309-76-9 Molecular Formula: C13H15NO2S Molecular Weight (g/mol): 249.33 Synonym: BAY 11-7085,(E)-3-[[4-(1,1-Dimethylethyl)phenyl]sulfonyl]-2-propenenitrile,BAY 117083 IUPAC Name: (E)-3-(4-tert-butylphenyl)sulfonylprop-2-enenitrile SMILES: CC(C)(C)c1ccc(cc1)S(=O)(=O)\C=C\C#N
2
Promotions Available
Tocris Bioscience™ Bay 11-7085
Tocris Bioscience™ Bay 11-7085
3
BAY 11-7085, MedChemExpress
MedChemExpress BAY 11-7085 (BAY 11-7083) is an inhibitor of NF-κB activation and phosphorylation of IκBα; it stabilizes IκBα with an IC50 of 10 μM.