CAS RN 195886-79-4
IUPAC Name:
2-(difluoromethyl)-1,4-difluorobenzene
Synonyms:
2-(difluoromethyl)-1,4-difluorobenzene
Molecular Weight (g/mol):
164.1
Molecular Formula:
C7H4F4
InChi Key:
FDUVCOVVYFCQEH-UHFFFAOYSA-N
SMILES:
FC(F)C1=C(F)C=CC(F)=C1
1
–
1
of
1
results
1
2-(Difluoromethyl)-1,4-difluorobenzene, 97%, Thermo Scientific™
CAS: 195886-79-4 Molecular Formula: C7H4F4 Molecular Weight (g/mol): 164.1 InChI Key: FDUVCOVVYFCQEH-UHFFFAOYSA-N Synonym: 2-difluoromethyl-1,4-difluorobenzene,2,5-difluorobenzal fluoride,1-difluoromethyl-2,5-difluorobenzene,2,4-difluoro-1-difluoromethyl benzene PubChem CID: 46311703 IUPAC Name: 2-(difluoromethyl)-1,4-difluorobenzene SMILES: C1=CC(=C(C=C1F)C(F)F)F