CAS RN 19574-66-4
CAS RN 19574-66-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
3-(Diphenylmethoxy)-1,2-propanediol, TRC
CAS: 19574-66-4 Molecular Formula: C16 H18 O3 Molecular Weight (g/mol): 258.31 Synonym: 3-(Diphenylmethoxy)-1,2-propanediol,3-(Benzhydryloxy)propane-1,2-diol IUPAC Name: 3-benzhydryloxypropane-1,2-diol SMILES: OCC(O)COC(c1ccccc1)c2ccccc2
2
3-(Benzhydryloxy)propane-1,2-diol, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.