missing translation for 'onlineSavingsMsg'
Learn More

3-(Benzhydryloxy)propane-1,2-diol, Mikromol™

Product Code. 30464070
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30464070 4 x 25 mg 100mg
30049787 25 mg 25mg
30050539 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30464070 Supplier Mikromol Supplier No. MM0490.0900254AMP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3-(Benzhydryloxy)propane-1,2-diol, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(benzhydryloxy)propane-1,2-diol
CAS 19574-66-4
Analyte or Component Names 3-(benzhydryloxy)propane-1,2-diol
Shipping Condition Room Temperature
Molecular Formula C16 H18 O3
Certifications/Compliance ISO 9001
Synonym 3-(Diphenylmethoxy)-1,2-propanediol, 3-(Benzhydryloxy)propane-1,2-diol, Diphenhydramine Related Compound B
InChI Formula InChI=1S/C16H18O3/c17-11-15(18)12-19-16(13-7-3-1-4-8-13)14-9-5-2-6-10-14/h1-10,15-18H,11-12H2
Solution Type Neat
SMILES OCC(O)COC(c1ccccc1)c2ccccc2
IUPAC Name 3-benzhydryloxypropane-1,2-diol
Molecular Weight (g/mol) 258.31
Quantity 4 x 25 mg
Formula Weight 258.1256 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.