CAS RN 19545-26-7
Wortmannin, 98%
CAS: 19545-26-7 Molecular Formula: C23H24O8 Molecular Weight (g/mol): 428.44 MDL Number: MFCD00133927 InChI Key: QDLHCMPXEPAAMD-QAIWCSMKSA-N Synonym: wortmannin,wartmannin,antibiotic sl-2052,unii-xva4o219qw,xva4o219qw,pi 3-kinase inhibitor,1alpha,11alpha-11-acetyloxy-1-methoxymethyl-2-oxaandrosta-5,8-dieno 6,5,4-bc furan-3,7,17-trione,1r,3r,5s,9r,18s-18-methoxymethyl-1,5-dimethyl-6,11,16-trioxo-13,17-dioxapentacyclo 10.6.1.0^ 2,10 .0^ 5,9 .0^ 15,19 nonadeca-2 10 ,12 19 ,14-trien-3-yl acetate,1s,6br,9as,11r,11br-9a,11b-dimethyl-1-methyloxy methyl-3,6,9-trioxo-1,6,6b,7,8,9,9a,10,11,11b-decahydro-3h-furo 4,3,2-de indeno 4,5-h isochromen-11-yl acetate,wortmanin PubChem CID: 312145 ChEBI: CHEBI:52289 IUPAC Name: (1R,3R,5S,9R,18S)-18-(methoxymethyl)-1,5-dimethyl-6,11,16-trioxo-13,17-dioxapentacyclo[10.6.1.0²,¹⁰.0⁵,⁹.0¹⁵,¹⁹]nonadeca-2(10),12(19),14-trien-3-yl acetate SMILES: COC[C@H]1OC(=O)C2=COC3=C2[C@@]1(C)C1=C([C@@H]2CCC(=O)[C@@]2(C)C[C@H]1OC(C)=O)C3=O
Wortmannin, >99% purity, For TLC, HPLC analysis, 97.96%, MP Biomedicals™
CAS: 19545-26-7 Molecular Formula: C23H24O8 Molecular Weight (g/mol): 428.44 MDL Number: MFCD00133927 InChI Key: QDLHCMPXEPAAMD-QAIWCSMKSA-N Synonym: wortmannin,wartmannin,antibiotic sl-2052,unii-xva4o219qw,xva4o219qw,pi 3-kinase inhibitor,1alpha,11alpha-11-acetyloxy-1-methoxymethyl-2-oxaandrosta-5,8-dieno 6,5,4-bc furan-3,7,17-trione,1r,3r,5s,9r,18s-18-methoxymethyl-1,5-dimethyl-6,11,16-trioxo-13,17-dioxapentacyclo 10.6.1.0^ 2,10 .0^ 5,9 .0^ 15,19 nonadeca-2 10 ,12 19 ,14-trien-3-yl acetate,1s,6br,9as,11r,11br-9a,11b-dimethyl-1-methyloxy methyl-3,6,9-trioxo-1,6,6b,7,8,9,9a,10,11,11b-decahydro-3h-furo 4,3,2-de indeno 4,5-h isochromen-11-yl acetate,wortmanin PubChem CID: 312145 ChEBI: CHEBI:52289 IUPAC Name: (1R,3R,5S,9R,18S)-18-(methoxymethyl)-1,5-dimethyl-6,11,16-trioxo-13,17-dioxapentacyclo[10.6.1.0²,¹⁰.0⁵,⁹.0¹⁵,¹⁹]nonadeca-2(10),12(19),14-trien-3-yl acetate SMILES: COC[C@H]1OC(=O)C2=COC3=C2[C@@]1(C)C1=C([C@@H]2CCC(=O)[C@@]2(C)C[C@H]1OC(C)=O)C3=O
R&D Systems™ Wortmannin
CAS: 19545-26-7 Molecular Formula: C23H24O8
Wortmannin, TRC
CAS: 19545-26-7 Molecular Formula: C23H24O8 Molecular Weight (g/mol): 428.43 Synonym: (+)-Wortmannin,KY 12420,[1S-(1α,6bα,9aβ,11α,11bβ)]-11-(Acetyloxy)-1,6b,7,8,9a,10,11,11b-octahydro-1-(methoxymethyl)-9a,11b-dimethyl-3H-Furo[4,3,2-de]indeno[4,5-h]-2-benzopyran-3,6,9-trione,(1S,6bR,9aS,11R,11bR)-11-(Acetyloxy)-1,6b,7,8,9a,10,11,11b-octahydro-1-(methoxymethyl)-9a,11b-dimethyl-3H-furo[4,3,2-de]indeno[4,5-h]-2-benzopyran-3,6,9-trione,(1S,6bR,9aS,11R,11bR)-1-(Methoxymethyl)-9a,11b-dimethyl-3,6,9-trioxo-3,6,6b,7,8,9,9a,10,11,11b-decahydro-1H-furo[4,3,2-de]indeno[4,5-h]isochromen-11-yl Acetate SMILES: COC[C@H]1OC(=O)c2coc3C(=O)C4=C([C@@H](C[C@@]5(C)[C@H]4CCC5=O)OC(=O)C)[C@]1(C)c23
Wortmannin, MedChemExpress
MedChemExpress Wortmannin (SL-2052; KY-12420) is a potent, selective and irreversible PI3K inhibitor with an IC50 of 3 nM. Wortmannin also blocks autophagy formation, and potently inhibits Polo-like kinase 1 (PlK1) and Plk3 with IC50s of 5.8 and 48 nM, respectively.
Thermo Scientific Chemicals Wortmannin, Penicillium funiculosum, 99+%
CAS: 19545-26-7 Molecular Formula: C23H24O8 Synonym: KY 12420