accessibility menu, dialog, popup

UserName

CAS RN 19485-35-9

CH3NNNN

CAS RN 19485-35-9

IUPAC Name: 1-methyl-1H-imidazole-4,5-dicarbonitrile
Synonyms: 1-methylimidazole-4,5-dicarbonitrile
Molecular Weight (g/mol): 132.13
Molecular Formula: C6H4N4
InChi Key: SUOVGHDPXJUFME-UHFFFAOYSA-N
SMILES: CN1C=NC(C#N)=C1C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results