Learn More
1-Methylimidazole-4,5-dicarbonitrile, 97%, Thermo Scientific™
Product Code. 11496526
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11496526
Brand: Thermo Scientific Alfa Aesar B24786.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 19485-35-9
Molecular Formula C6H4N4
Molecular Weight (g/mol) 132.13
MDL Number MFCD01567299
InChI Key SUOVGHDPXJUFME-UHFFFAOYSA-N
PubChem CID 583268
IUPAC Name 1-methylimidazole-4,5-dicarbonitrile
SMILES CN1C=NC(C#N)=C1C#N
Specifications
Melting Point 83°C to 86°C
Quantity 25 g
UN Number UN3439
Synonym 1-methyl-1h-imidazole-4,5-dicarbonitrile, 4,5-dicyano-1-methyl-1h-imidazole, 4,5-dicyano-1-methylimidazole, imidazole, 4,5-dicyano-1-methyl, 1h-imidazole-4,5-dicarbonitrile, 1-methyl, pubchem8999, acmc-1bvri, 1-methyl-4,5-dicyano-1h-imidazole, 1h-imidazole-4,5-dicarbonitrile,1-methyl, 1-methyl-1h-imidazole-4,5-dicarbonitrile #
Formula Weight 132.14
Percent Purity 97%
Chemical Name or Material 1-Methylimidazole-4,5-dicarbonitrile