accessibility menu, dialog, popup

UserName

FClFO

CAS RN 190011-87-1

IUPAC Name: 3-chloro-2,6-difluorobenzaldehyde
Synonyms: 3-chloro-2,6-difluorobenzaldehyde
Molecular Weight (g/mol): 176.55
Molecular Formula: C7H3ClF2O
InChi Key: HFKZZEDGXXYRDW-UHFFFAOYSA-N
SMILES: FC1=CC=C(Cl)C(F)=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results