accessibility menu, dialog, popup

UserName

CH3SOOH

CAS RN 18698-73-2

IUPAC Name: 2-[3-(methylsulfanyl)phenyl]acetic acid
Synonyms: [3-(methylsulfanyl)phenyl]acetic acid
Molecular Weight (g/mol): 182.24
Molecular Formula: C9H10O2S
InChi Key: AKYCOAMUNLESIH-UHFFFAOYSA-N
SMILES: CSC1=CC=CC(CC(O)=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results