accessibility menu, dialog, popup

UserName

CH3SiH3C

CAS RN 18666-26-7

IUPAC Name: bis(1H-inden-1-yl)dimethylsilane
Synonyms: bis(1H-inden-1-yl)dimethylsilane
Molecular Weight (g/mol): 288.47
Molecular Formula: C20H20Si
InChi Key: DXYTUIWIDDBVLU-UHFFFAOYNA-N
SMILES: C[Si](C)(C1C=CC2=CC=CC=C12)C1C=CC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)