CAS RN 185413-30-3
CAS RN 185413-30-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
AF 12198, TRC
CAS: 185413-30-3 Synonym: N-Acetyl-L-phenylalanyl-L-α-glutamyl-L-tryptophyl-L-threonyl-L-prolylglycyl-L-tryptophyl-L-tyrosyl-L-glutaminyl-(2S)-2-azetidinecarbonyl-L-tyrosyl-L-alanyl-L-leucyl-L-prolyl-L-Leucinamide∣AF 12198