Learn More
AF 12198, TRC
Product Code. 17982195
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17982195 25 mg 25mg
17924265 5 mg 5mg
17974605 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17982195 Supplier Toronto Research Chemicals Supplier No. TRCA35502025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 185413-30-3
Synonym N-Acetyl-L-phenylalanyl-L-α-glutamyl-L-tryptophyl-L-threonyl-L-prolylglycyl-L-tryptophyl-L-tyrosyl-L-glutaminyl-(2S)-2-azetidinecarbonyl-L-tyrosyl-L-alanyl-L-leucyl-L-prolyl-L-Leucinamide∣AF 12198
Tekniske data
Format Neat
Molecular Formula C96H123N19O22
Molecular Weight (g/mol) 1895.12, 1895.15
InChI Key VASLMBMCJADVGR-UHFFFAOYSA-N
SMILES CC(C)CC(NC(=O)C1CCCN1C(=O)C(CC(C)C)NC(=O)C(C)NC(=O)C(CC1=CC=C(O)C=C1)NC(=O)C1CCN1C(=O)C(CCC(N)=O)NC(=O)C(CC1=CC=C(O)C=C1)NC(=O)C(CC1=CNC2=CC=CC=C12)NC(=O)CNC(=O)C1CCCN1C(=O)C(NC(=O)C(CC1=CNC2=CC=CC=C12)NC(=O)C(CCC(O)=O)NC(=O)C(CC1=CC=CC=C1)NC(C)=O)C(C)O)C(N)=O
IUPAC Name 4-[(1-{[1-(2-{[({1-[(1-{[4-carbamoyl-1-(2-{[1-({1-[(1-{2-[(1-carbamoyl-3-methylbutyl)carbamoyl]pyrrolidin-1-yl}-4-methyl-1-oxopentan-2-yl)carbamoyl]ethyl}carbamoyl)-2-(4-hydroxyphenyl)ethyl]carbamoyl}azetidin-1-yl)-1-oxobutan-2-yl]carbamoyl}-2-(4-hydroxyphenyl)ethyl)carbamoyl]-2-(1H-indol-3-yl)ethyl}carbamoyl)methyl]carbamoyl}pyrrolidin-1-yl)-3-hydroxy-1-oxobutan-2-yl]carbamoyl}-2-(1H-indol-3-yl)ethyl)carbamoyl]-4-(2-acetamido-3-phenylpropanamido)butanoic acid
Quantity 25 mg
Shipping Condition Room Temperature
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.