CAS RN 1797133-13-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Trityl-deshydroxymethyl Losartan, TRC
CAS: 1797133-13-1 Molecular Formula: C40H35ClN6 Molecular Weight (g/mol): 635.21 Synonym: 5-[4'-[(2-Butyl-4-chloro-1H-imidazol-1-yl)methyl][1,1'-biphenyl]-2-yl]-2-(triphenylmethyl)-2H-tetrazole IUPAC Name: 5-[2-[4-[(2-butyl-4-chloroimidazol-1-yl)methyl]phenyl]phenyl]-2-trityltetrazole SMILES: CCCCc1nc(Cl)cn1Cc2ccc(cc2)c3ccccc3c4nnn(n4)C(c5ccccc5)(c6ccccc6)c7ccccc7