accessibility menu, dialog, popup

UserName

OPOOClClCl

CAS RN 17776-78-2

IUPAC Name: bis(2-chlorophenyl) phosphorochloridate
Synonyms: bis(2-chlorophenyl) phosphorochloridate
Molecular Weight (g/mol): 337.52
Molecular Formula: C12H8Cl3O3P
InChi Key: ZLSGEMKKBUVQOM-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1OP(Cl)(=O)OC1=CC=CC=C1Cl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results