accessibility menu, dialog, popup

UserName

H3COOH(E/Z)

CAS RN 17735-94-3

IUPAC Name: icos-13-enoic acid
Synonyms: paullinic acid
Molecular Weight (g/mol): 310.52
Molecular Formula: C20H38O2
InChi Key: URXZXNYJPAJJOQ-UHFFFAOYSA-N
SMILES: CCCCCCC=CCCCCCCCCCCCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results