missing translation for 'onlineSavingsMsg'
Learn More

Paullinic acid, MedChemExpress

Product Code. 16474748
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16474748 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16474748 Supplier Medchem Express Supplier No. HY113094

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Paullinic acid is a long-chain fatty acid that has been detected in multiple biofluids, such as blood and urine.

Paullinic acid is a long-chain fatty acid that has been detected in multiple biofluids, such as blood and urine.

Specifications

Chemical Name or Material Paullinic acid
CAS 17735-94-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H38O2
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (322.05 mM; Need ultrasonic)
SMILES CCCCCC/C=C\CCCCCCCCCCCC(O)=O
Molecular Weight (g/mol) 310.51
Formula Weight 310.51
Percent Purity 97.0%
Grade Research
Physical Form Oil
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.