CAS RN 176957-55-4
CAS RN 176957-55-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
CCT007093, MedChemExpress
MedChemExpress CCT007093 is an effective protein phosphatase 1D (PPM1D Wip1) inhibitor. Wip1 inhibition can activate the mTORC1 pathway and enhance hepatocyte proliferation after hepatectomy.
2
PPM1D Phosphatase Inhibitor, Thermo Scientific™
Acts as a PPM1D inhibitor