missing translation for 'onlineSavingsMsg'
Learn More

CCT007093, MedChemExpress

Product Code. 16412957
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16412957

Brand: Medchem Express HY15880

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CCT007093 is an effective protein phosphatase 1D (PPM1D Wip1) inhibitor. Wip1 inhibition can activate the mTORC1 pathway and enhance hepatocyte proliferation after hepatectomy.

CCT007093 is an effective protein phosphatase 1D (PPM1D Wip1) inhibitor. Wip1 inhibition can activate the mTORC1 pathway and enhance hepatocyte proliferation after hepatectomy.

Specifications

Chemical Name or Material CCT007093
Physical Form Solid
CAS 176957-55-4
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C15H12OS2
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMF : 3.33 mg/mL (12.23 mM; Need ultrasonic) ∣DMSO : ≥ 2.8 mg/mL (10.28 mM)
SMILES O=C1/C(CC/C1=C\C2=CC=CS2)=C/C3=CC=CS3
Molecular Weight (g/mol) 272.39
Formula Weight 272.39
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.