accessibility menu, dialog, popup

UserName

CAS RN 175278-15-6

IUPAC Name: 4-bromo-2-(trifluoromethoxy)benzene-1-thiol
Synonyms: 4-bromo-2-(trifluoromethoxy)benzenethiol
Molecular Weight (g/mol): 273.07
Molecular Formula: C7H4BrF3OS
InChi Key: WWZNHDMEBZHFTK-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=C(S)C=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results