accessibility menu, dialog, popup

UserName

BrOFFFBr

CAS RN 175278-13-4

IUPAC Name: 1,4-dibromo-2-(trifluoromethoxy)benzene
Synonyms: 1,4-dibromo-2-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 319.9
Molecular Formula: C7H3Br2F3O
InChi Key: WVMFAEDJOGXROT-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=C(Br)C=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results