CAS RN 175278-09-8
CAS RN 175278-09-8
IUPAC Name:
4-bromo-2-(trifluoromethoxy)aniline
Synonyms:
4-bromo-2-(trifluoromethoxy)aniline
Molecular Weight (g/mol):
256.02
Molecular Formula:
C7H5BrF3NO
InChi Key:
QVILSWLYJYMGRN-UHFFFAOYSA-N
SMILES:
NC1=CC=C(Br)C=C1OC(F)(F)F
1
–
2
of
2
results
2
4-Bromo-2-(trifluoromethoxy)aniline, 97%, Thermo Scientific™
CAS: 175278-09-8 Molecular Formula: C7H5BrF3NO Molecular Weight (g/mol): 256.02 MDL Number: MFCD00179338 InChI Key: QVILSWLYJYMGRN-UHFFFAOYSA-N Synonym: 4-bromo-2-trifluoromethoxy aniline,4-bromo-2-trifluoromethoxy-phenylamine,2-amino-5-bromo trifluoromethoxy benzene,benzenamine, 4-bromo-2-trifluoromethoxy,1-amino-4-bromo-2-trifluoromethoxy benzene,buttpark 50\01-74,4-bromo-2-trifluoromethoxy phenylamine,pubchem2930,acmc-1c9j8,ksc184e5f PubChem CID: 737382 IUPAC Name: 4-bromo-2-(trifluoromethoxy)aniline SMILES: NC1=CC=C(Br)C=C1OC(F)(F)F