accessibility menu, dialog, popup

UserName

NNHNClCl

CAS RN 175277-80-2

IUPAC Name: 1-(3,5-dichloropyridin-4-yl)piperazine
Synonyms: 1-(3,5-dichloropyridin-4-yl)piperazine
Molecular Weight (g/mol): 232.11
Molecular Formula: C9H11Cl2N3
InChi Key: GPCFTLKHUONISF-UHFFFAOYSA-N
SMILES: ClC1=CN=CC(Cl)=C1N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results