CAS RN 1748-81-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Carabron, TRC
CAS: 1748-81-8 Molecular Formula: C15 H20 O3 Molecular Weight (g/mol): 248.317 Synonym: (3aR,4aS,5S,5aR,6aR)-Octahydro-5a-methyl-3-methylene-5-(3-oxobutyl)-2H-cyclopropa[f]benzofuran-2-one,Carabrone,Caropesia Lactone IUPAC Name: (3aR,4aS,5S,5aR,6aR)-5a-methyl-3-methylidene-5-(3-oxobutyl)-3a,4,4a,5,6,6a-hexahydrocyclopropa[f][1]benzofuran-2-one SMILES: CC(=O)CC[C@H]1[C@@H]2C[C@H]3[C@@H](C[C@]12C)OC(=O)C3=C