accessibility menu, dialog, popup

UserName

CAS RN 17363-02-9

IUPAC Name: platinum(4+) diammonium hexabromide
Synonyms: platinum(4+) diammonium hexabromide
Molecular Weight (g/mol): 710.59
Molecular Formula: Br6H8N2Pt
InChi Key: YLDMEZNZYLWNNL-UHFFFAOYSA-J
SMILES: [NH4+].[NH4+].[Br-].[Br-].[Br-].[Br-].[Br-].[Br-].[Pt+4]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results