accessibility menu, dialog, popup

UserName

NH2OOH

CAS RN 17194-82-0

IUPAC Name: 2-(4-hydroxyphenyl)acetamide
Synonyms: P-hydroxy-phenylacetamide
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: YBPAYPRLUDCSEY-UHFFFAOYSA-N
SMILES: NC(=O)CC1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (5)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (4)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (1)