CAS RN 165377-44-6
CAS RN 165377-44-6
IUPAC Name:
1-[2-(3,4-dimethoxyphenyl)ethyl]-4-(3-phenylpropyl)piperazine dihydrochloride
Synonyms:
cutamesine dihydrochloride
Molecular Weight (g/mol):
441.44
Molecular Formula:
C23H34Cl2N2O2
InChi Key:
XWOXAKBQEMQMFH-UHFFFAOYSA-N
SMILES:
Cl.Cl.COC1=C(OC)C=C(CCN2CCN(CCCC3=CC=CC=C3)CC2)C=C1
1
–
2
of
2
results
1
Tocris Bioscience™ SA 4503 dihydrochloride
CAS: 165377-44-6 Molecular Formula: C23H34Cl2N2O2
2
Cutamesine dihydrochloride, MedChemExpress
MedChemExpress Cutamesine dihydrochloride (SA4503 dihydrochloride; AGY94806 dihydrochloride) is a potent Sigma 1 receptor agonist with an IC50 of 17.4 nM in guinea pig brain membranes.