CAS RN 163521-19-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Vilazodone Carboxylic Acid, TRC
CAS: 163521-19-5 Molecular Formula: C26 H26 N4 O3 Molecular Weight (g/mol): 442.51 Synonym: 2-Benzofurancarboxylic acid, 5-[4-[4-(5-cyano-1H-indol-3-yl)butyl]-1-piperazinyl]-,5-[4-[4-(5-Cyano-1H-indol-3-yl)butyl]-1-piperazinyl]-2-benzofurancarboxylic acid,Vilazodone carboxylic acid,5-[4-[4-(5-Cyano-1H-indol-3-yl)butyl]piperazin-1-yl]benzofuran-2-carboxylic acid IUPAC Name: 5-[4-[4-(5-cyano-1H-indol-3-yl)butyl]piperazin-1-yl]-1-benzofuran-2-carboxylic acid SMILES: OC(=O)c1oc2ccc(cc2c1)N3CCN(CCCCc4c[nH]c5ccc(cc45)C#N)CC3