accessibility menu, dialog, popup

UserName

CAS RN 163428-90-8

CH3OOH3COO

CAS RN 163428-90-8

IUPAC Name: 2-(3,4-dimethoxyphenyl)-2-oxoacetaldehyde
Synonyms: 2-(3,4-dimethoxyphenyl)-2-oxoacetaldehyde
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: QQSUAMUBZNQYGE-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1OC)C(=O)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results