accessibility menu, dialog, popup

UserName

CH3NNOOH

CAS RN 16034-46-1

IUPAC Name: 1-methyl-1H-pyrazole-5-carboxylic acid
Synonyms: 2-methylpyrazole-3-carboxylic acid
Molecular Weight (g/mol): 126.12
Molecular Formula: C5H6N2O2
InChi Key: JREJQAWGQCMSIY-UHFFFAOYSA-N
SMILES: CN1N=CC=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results