accessibility menu, dialog, popup

UserName

CAS RN 15965-54-5

CH3ONHNCl

CAS RN 15965-54-5

IUPAC Name: 2-chloro-6-methoxy-1H-1,3-benzodiazole
Synonyms: 2-chloro-5-methoxy-3H-1,3-benzodiazole
Molecular Weight (g/mol): 182.61
Molecular Formula: C8H7ClN2O
InChi Key: FMDGYQOERIOABX-UHFFFAOYSA-N
SMILES: COC1=CC=C2N=C(Cl)NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results