accessibility menu, dialog, popup

UserName

CH3SOOHN

CAS RN 1576-37-0

IUPAC Name: N-benzyl-4-methylbenzene-1-sulfonamide
Synonyms: N-benzyl-4-methylbenzenesulfonamide
Molecular Weight (g/mol): 261.34
Molecular Formula: C14H15NO2S
InChi Key: WTHKAJZQYNKTCJ-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)NCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)